Would you like to log in again?
High Resolution Gold on Carbon Test, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
79510-04 79510-01 79510-02 79510-05 79510-03
100494-952EA 398.58 USD InStock
100494-952 100494-960 100494-958 100494-956 100494-954
High Resolution Gold on Carbon Test, Electron Microscopy Sciences
Specimen Mounting Supplies
The gold on carbon specimen is the resolution test of SE and BSE imaging and also for chemical mapping in high-resolution systems such as an Auger scanning instrument.
  • Gold particles range from 5 to 150 nm approximately
  • Each test specimen has a square grid pattern with large crystals in the center of each square
  • Very fine crystals at the edges of each grid
Thus medium and high resolution gap tests are performed on the same specimen. Meantime, the larger crystals show facets which allow an assessment of the gray level reproduction available at high resolution.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty