Would you like to log in again?
Duplex Specimen, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
79521-01
100494-988EA 1506.86 USD InStock
100494-988
Duplex Specimen, Electron Microscopy Sciences
Specimens Preserved Specimens Display Specimens
An alternative and very sensitive test is by means of an alloy with two major Copper/Zinc phases separated by an atomic number difference of 0.1. The light phase illustrated in the micrograph has a mean atomic number of 29.47 and the dark phase a mean atomic number of 29.37.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty