Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
MAC X-Ray Microanalysis Individual Standards, Micro-Analysis Consultants
Supplier: Electron Microscopy Sciences
80074-CU 80074-PD 80074-BAS 80074-SC 80074-RE 80074-SB 80074-MG 80074-MO 80074-HF 80074-C 80074-Y 80074-GD 80074-NBS-SS 80074-IR 80074-NBS-CA 80074-NBS-SL 80074-CR 80074-BAS-FC 80074-BI 80074-SI 80074-BE 80074-NI 80074-NBS-HS 80074-AG 80074-GE 80074-AS 80074-RH 80074-MN 80074-NBS 80074-PT 80074-SE 80074-TA 80074-AU 80074-RU 80074-B 80074-AL 80074-TL 80074-FE 80074-TI 80074-BAS-S 80074-ZR 80074-TE 80074-SN 80074-NB 80074-ZN 80074-W 80074-CO 80074-V 80074-OS 80074-CD
100495-124EA 602.34 USD InStock
100495-124 102099-018 100495-126 102099-012 100495-042 100495-120 100495-044 102099-014 100495-122 100495-128 102099-020 100495-096 100495-112 102099-026 100495-036 100495-114 102099-028 100495-038 100495-076 102099-022 100495-110 100495-078 102099-024 100495-116 100495-118 100495-082 100495-040 100495-084 100495-080 100495-068 100495-102 100495-104 100495-064 100495-066 100495-100 100495-106 100495-108 100495-072 100495-074 100495-070 100495-134 100495-058 100495-130 100495-098 100495-054 100495-132 100495-056 100495-060 102099-010 100495-062
MAC X-Ray Microanalysis Individual Standards, Micro-Analysis Consultants
Standards Color Reference and Spectroscopy Standards
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems.
All the standards are polished to a 0.25 micron diamond finish and carbon coated. Supplied in 2, 3, and 5 mm diameter and a location map for standard identification. They are supplied with fully authenticated certificates of analysis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty