Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
MAC X-ray Microanalysis Individual Standards, Grains, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
80075-B 80075-HF 80075-C 80075-BAS-HA 80075-AU 80075-RU 80075-OS 80075-MN 80075-BAS-FC 80075-SN 80075-TI 80075-IR 80075-SC 80075-ZR 80075-NI 80075-NBS-710 80075-CR 80075-CU 80075-TE 80075-AL 80075-MO 80075-RH 80075-MG 80075-SB 80075-V 80075-NBS-316 80075-ZN 80075-GD 80075-GE 80075-AG 80075-BE 80075-Y 80075-TL 80075-PT 80075-W 80075-NB 80075-BI 80075-FE 80075-TA 80075-SI 80075-BAS-G 80075-BAS 80075-CD 80075-NBS-478 80075-NBS 80075-RE 80075-CO 80075-SE 80075-NBS-SRM 80075-PD 80075-AS
100495-200EA 487.03 USD InStock
100495-200 100495-168 100495-202 100495-164 100495-166 100495-208 100495-204 100495-206 100495-170 100495-172 100495-156 100495-234 100495-158 100495-236 100495-196 100495-230 100495-198 100495-154 100495-232 102099-030 100495-160 102099-032 100495-162 100495-222 102099-038 100495-224 102099-034 100495-142 102099-036 100495-220 100495-144 100495-226 100495-228 102099-040 102099-042 100495-178 100495-212 100495-136 100495-214 100495-174 102099-044 100495-176 100495-210 102099-046 100495-138 100495-216 100495-218 100495-182 100495-184 100495-140 100495-180
MAC X-ray Microanalysis Individual Standards, Grains, Electron Microscopy Sciences
Standards Color Reference and Spectroscopy Standards
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems.
All the standards are polished to a 0.25 micron diamond finish and carbon coated. They are supplied with a location map for standard identification. Grains of standard materials are available and they measure approximately 2 x 2 mm in size. They are supplied with fully authenticated certificates of analysis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty