Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
MAC Thin Film Standards for TEM, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
80080-20 80080-10 80080-01 80080-25 80080-15 80080-05 80080-50 80080-30 80080-40
100495-244EA 347.88 USD InStock
100495-244 100495-256 100495-246 100495-258 100495-252 100495-254 100495-260 100495-250 100495-248
MAC Thin Film Standards for TEM, Electron Microscopy Sciences
Standards Color Reference and Spectroscopy Standards
These standards are supplied as fine powders that are dispersed onto holey-carbon films and are selected from the range of certified natural and synthetic compounds already in use to prepare bulk analysis standards.
  • The holey-carbon films are supported by a 3.05 mm copper grid (400 mesh)
The Universal set of Thin Film Standards are Ag2Te, BaSO4, Be3Al2Si6O18, Bi2Se3, CaMoO4, CaWO4, CdTe, CeAl2, Cu2S, FeCr2O4, GaAs, Gd3Ga5O12, HgTe, InP, InSb, KAlSi3O8, LaB6, Li2Ta2O6, (Mg,Fe)2SiO4, Na3AlF6, Pb-Ge glass, SrTiO3, TlBr, ZnS, ZrSiO4.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty