Would you like to log in again?
SEM, Finder Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
80100-CU 80100-NI
100495-320EA 347.89 USD InStock
100495-320 100495-318
SEM, Finder Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
Finder grids for SEM; a valuable tool for analytical studies. The grids are available in 10 mm diameter and they can be placed directly on the stub surface and used to identify the area of interest in which the studies are performed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty