Would you like to log in again?
Light Pipe Scintillators, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
82001-100 82000-89 82000-74 82000-104 82001-88 82000-X 82001-120 82000-10 82001-X 82008-RS 82004-60 82005-60
100495-420EA 1333.89 USD InStock
100495-420 100495-410 100495-422 100495-412 102099-374 100502-322 100495-418 100502-324 100495-424 100495-414 100502-320 100495-416
Light Pipe Scintillators, Electron Microscopy Sciences
Lights Microscope Bulbs
The light pipe is the connection between the scintillator and the phosphomultiplier tube (PMT). It transfers the photons produced by the scintillator for detection by the PMT.
  • Scintillator material coated directly on the light pipe which improves the signal
  • Re-coating is typically needed twice per year
Light Pipes are available in optical quality quartz and optical grade acrylic. Cambridge reduced-tip models are only available in acrylic.
Delivery information: Includes one Re-coating Certificate. High voltage wire and corona ring are included, JEOL pipes include a flange.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty