Would you like to log in again?
Embra Oval Slot Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
E0420-AL E2010-SS E0420-SS E0420-MO E0420-TI E2010-AL E2010-TI E2010-MO
100495-530EA 107.33 USD InStock
100495-530 100495-552 100495-532 100495-554 100495-528 100495-556 100495-558 100495-526
Embra Oval Slot Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
EMBRA electroformed grids combine a high open area with a rigid construction.
  • Measures 3.05 mm in diameter
  • Thickness for oval hole grids is about 50 micron
  • Relatively easy handling
Available in Copper (Cu), Nickel (Ni), Gold (Au), Stainless Steel (SS), Titanium (Ti), Molybdenum (Mo) and Aluminum (Al).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty