Would you like to log in again?
Embra Square Mesh, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
E100-AL E300-TI E200-SS E100-SS E300-MO E300-AL E200-MO E200-TI E100-MO E200-AL E100-TI
100495-564EA 260.21 USD InStock
100495-564 100495-542 100495-544 100495-560 100495-550 100495-562 100495-540 100495-538 100495-534 100495-536 100495-548
Embra Square Mesh, Electron Microscopy Sciences
Graticules, Reticles and Grids
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. Measures 3.05 mm in diameter and 16 micron in thickness. Material: Copper (Cu), Nickel (Ni), Gold (Au), Stainless Steel (SS), Titanium (Ti), Molybdenum (Mo), Aluminum (Al).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty