Would you like to log in again?
Embra Finder Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
EF400-AU EF200-CU EF300-NI EF100-NI EF100-CU EF300-AU EF400-NI EF200-NI EF400-CU EF300-CU EF200-AU
100495-596EA 122.87 USD InStock
100495-596 100495-586 100495-598 100495-588 100495-592 100495-594 100495-604 100502-982 100495-590 100495-600 100495-602
Embra Finder Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
Standard 3.05 mm diameter grids which have one straight and one round cut out from the rim which assists in the orientation of the grid.
Ordering information: Available in Copper, Nickel, and Gold Grids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty