Would you like to log in again?
Finder Grids for Electron Microscopy, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
G300F1-AU G200F1-CU G400F1-NI G300F1-NI G200F2-CP G200F2-NI G200F1-NI G200F1-CP G200F1-AU G400F1-AU G200F2-CU G200HF3-CU G400F1-CU G200F2-AU G300F1-CU G200HF3-NI
100495-640EA 101.07 USD InStock
100495-640 103303-102 100495-642 103303-104 100495-650 100495-648 100495-638 103302-936 100495-644 100495-646 103302-934 100502-338 102101-128 103302-156 103302-154 102101-130
Finder Grids for Electron Microscopy, Electron Microscopy Sciences
Graticules, Reticles and Grids
Alphabetical and numerical systems for recording specimen locations on the grid.
  • Copper, copper-paladium, nickel, molybdenum, and gold grids
  • 200, 300 and 400 mesh grids
  • Thick and thin bar grid patterns
Used to identify specific areas of mesh by an alphabetical or numerical code built into the mesh. 3.05 mm diameter, 0.7 mil (18 µm) thickness. G200F1 series has thick bars dividing regions into six thin bar areas, which are identified by a numeric system.
G200F2 series has thick bars dividing regions into nine thin bar areas, which are identified by alphabetical letters located in the center of the grid. G200HF3 series has 322 grids squares and each can be identified by reference to its unique combination of base two binary number and alphabet symbol (A-T). Zero is a short rounded solid pillar and 1 is a longer rounded solid pillar.
Bar size: 35 – 12 µm
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty