Would you like to log in again?
Alpha Numeric Index Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
G200F4-CP G200F4-NI G200F4-CU G200F4-AU
100495-652EA 125.25 USD InStock
100495-652 100495-654 100502-340 100495-656
Alpha Numeric Index Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
By employing a rectangular mesh the support value of the grid has been increased, offering a value intermediate between the most commonly used grid (200 Lines/") and (300 Lines/"). Each grid rectangle is asymmetrical having different outlines in all four corners. This allows for the orientation of the grid to be determined at microscopic levels.
The index feature enables the position of each grid to be identified with reference to the letters A-O along the horizontal axis and the numbers 1-15 along the vertical axis. The logo in the rim allows for precise orientation and aids in the identification of each side. Grids are available in Copper, Copper/Palladium, Nickel, and Gold. Corner Outline with Reference to Logo in the Rim.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty