Gilder aperture grids with a single slot.
- Copper, nickel, molybdenum, and gold grids
- Holes sizes of 50, 75, 100, 150, 200, 300, 400, 600, 800, 1000, 1500, and 2000 microns
- 50 micron thickness
Copper, nickel, molybdenum, and gold grids
Holes sizes of 50, 75, 100, 150, 200, 300, 400, 600, 800, 1000, 1500, and 2000 microns
50 micron thickness
Single Slot aperture TEM grids have a thickness of are 50 microns. Each grid is individually inspected.