Would you like to log in again?
Veco Handle Grids Hexagonal Mesh, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
HD300H-CU HD300H-NI HD400H-CU HD100H-CU HD200H-NI HD150H-NI HD150H-CU HD200H-CU HD100H-NI HD400H-NI
100495-860EA 49.11 USD InStock
100495-860 100495-850 100495-862 100495-852 100495-846 100495-868 100495-836 100495-866 100495-844 100495-834
Veco Handle Grids Hexagonal Mesh, Electron Microscopy Sciences
Graticules Reticles and Grids
Veco Grids offer superior handling characteristics
Veco Grids are the most rigid grids available. Measures 3.05 mm in diameter. Material: Copper (Cu); Nickel (Ni).
Plus, with a 0.8 mil thickness.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty