Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Micrometer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62061 62060
100496-130EA 72.23 USD InStock
100496-130 100496-128
Micrometer, Electron Microscopy Sciences
Calipers
Metric and English scale micrometers with friction stop; comes in a padded plastic case.
Model A: Calibrated to read 0 to 25 mm, in 0.01 mm divisions.
Model B: Calibrated to read 0 to 1", in 0.001" divisions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty