Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vernier Caliper, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
62064
100496-132EA 25.29 USD InStock
100496-132
Vernier Caliper, Electron Microscopy Sciences
Calipers
Double scale vernier caliper. Readable on the vernier scale to 1/128" and/or to 0.1 mm.

  • Nickel plated scale
  • Roller adjustment

Inch Scale: Reading from 0 to 5" in 1/16" divisions
Metric Scale: Reading from 0 to 120 mm in 1 mm divisions

Packaging: Packed in a plastic case.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty