Would you like to log in again?
Cleaning Basket, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62110
100496-146EA 10.51 USD InStock
100496-146
Cleaning Basket, Electron Microscopy Sciences
Baskets Wire Baskets
Made from stainless steel. Keeps small parts together when cleaning ultrasonically. A spring handle applies firm pressure to the basket to keep it closed when in use. Its overall length is 6". Spherical basket has a 38 mm (1 1/2") diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty