Would you like to log in again?
Angle Shield, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64464
100496-226EA 658.9 USD InStock
100496-226
Angle Shield, Electron Microscopy Sciences
Shields Safety Shields
Splash protecting in standing or sitting position provides good protection personal from pipetting or pouring chemicals.
  • Acrylic base work area is 50.8×30.5 cm (20×12") deep
  • Face is 50.9×38 cm (20×15") high
An extra wide, angled viewing area and low bend allow the shield to be used from a standing or sitting position. Made from 6.4 mm (¼") thick clear acrylic with non-skid feet for stability.
Caution: Not recommended for Beta use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty