Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mirror Stereo Viewer, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
68105
100496-270EA 989.88 USD InStock
100496-270
Mirror Stereo Viewer, Electron Microscopy Sciences
Magnifiers
For the viewing of 4"×5" (100×125 mm) stereo pairs. It stands on 4 folding legs. One of the legs is equipped with a length adjusting device, which provides stabilization to the unit during observation. The magnifier can be replaced depending on the application.

  • Base line spacing: 260 mm
  • Field of view without magnifier: 260×200 mm
  • Field of view with magnifier: 210×150 mm

Maximum size set up: 713×267×255 mm, net weight: 2.65 kg

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty