Would you like to log in again?
Duke Standards™ Particle-Size Standards, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70894 70895 70887 70898 70892 70889 70896 70891 70897 70883 70888 70890 70893 70886 70881 70885
100496-292EA 750.33 USD InStock
100496-292 100496-282 100496-290 100496-280 100496-296 100496-286 100496-294 100496-284 100496-300 100496-298 100496-288 100496-310 100496-304 100496-302 100496-308 100496-306
Duke Standards™ Particle-Size Standards, Electron Microscopy Sciences
Standards Chromatography Standards
Particle-size standards are certified for mean diameter and are traceable to the National Bureau of Standards (NBS). Duke Scientific's new line of standards in the submicrometer size range is called Nanosphere Size Standards™, and monosized polymer microspheres.
Nanosphere Size Standards are highly uniform polystyrene spheres calibrated in billionths of a meter (nanometer) with NBS traceable methodology. Nanosphere size standards are used in the calibration of electron microscopes, aerosol and liquid particle studies, chromatography columns, and laser light scattering studies. The 20 to 1000 nm range is suitable for checking the size of bacteria, viruses, ribosomes and sub-cellular components.
Monosized polymer microspheres are the most uniform spheres available. For the highest level of measurement accuracy in the 1 to 40 micron size, this series of size standards is recommended.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty