Would you like to log in again?
Micro Cleaning Solution, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71057 71056 71054 71055
100496-312EA 384.01 USD InStock
100496-312 100488-218 100496-314 100491-402
Micro Cleaning Solution, Electron Microscopy Sciences
Cleaning Agents Detergents and Surface Cleansers
Specially designed for ultrasonic cleaning machines. Cleans glass, most metals, plastics, and rubber.These can work with any concentration.
  • Biodegradable
  • Contains no phosphates
  • Removes radioactive contaminants
Not to be used on zinc, magnesium, or aluminum.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty