Would you like to log in again?
Slide Coating Adhesive, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
71302
100496-322EA 44.53 USD InStock
100496-322
Slide Coating Adhesive, Electron Microscopy Science
Stains Microscopy and staining reagents
Prepared from 3-aminopropyltriethoxysilane in acetone. Good for coating glass slides to adhere paraffin sections for further processing without sections falling off from the slide. READY TO USE.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty