Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Compact Slide Box for 25 Slides, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71461 71460
100501-096BG 25.94 USD InStock
100501-096 100496-324
Compact Slide Box for 25 Slides, Electron Microscopy Sciences
Microscope Slide Boxes
Made from durable polystyrene (with heavy walls).
Same characteristics as the Secured Hinge Microscope Slide Boxes, but with a slip-on cover. Holds 25 1x3" slides. Measures H x W x L: 1/4 x 3 1/2 x 5 1/2".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty