Would you like to log in again?
Sorensen's Phosphate Buffer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
11600-10 11682-10 11600-05 11601-10 11682-05
100496-400EA 65.11 USD InStock
100496-400 100496-398 100496-404 100496-402 102091-772
Sorensen's Phosphate Buffer, Electron Microscopy Sciences
Buffers Cell Culture Buffers
Sorensen's Phosphate Buffer is also know as phosphate mixed sodium salts. The osmolarity of a 0.1 M at a pH 7.2 is 226 mosmols; with the addition of 0.18 M of sucrose to the solution, the osmolarity raises to 425 mosmols.
  • pH can be specified when ordering
  • Maximum shelf life after receipt is 3 months
Ordering information: Certificates of analysis are available by lot or product number.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty