Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Earle’s balanced salt solutions (EBSS) Formula I
Supplier: Electron Microscopy Sciences
11765-05 11765-10
100496-414EA 54 USD InStock
100496-414 100496-412
Earle’s balanced salt solutions (EBSS) Formula I
Buffers Cell Culture Buffers
All of our Earle’s Balanced Salt Solution contains: Potassium Chloride, Sodium Chloride, Sodium Phosphate Monobasic, and D-glucose. Some formulations may contain additional components. All of the solutions are prepared 1X strength.
Earle’s Balanced Salt Solutions Formula I contains:
KCl: 0.157 M
NaCl: 0.157 M
NaH₂PO₄·H₂O: 0.157 M
MgSO₄·7H₂O: 0.157 M
CaCl₂,·2H₂O: 0.157 M
NaHCO₃: 0.157 M
D-Glusose: 1 g/L
Phenol Red: none
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty