Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Earle’s balanced salt solutions (EBSS) Formula II

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

11766-05 11766-10
100496-418EA 47.89 USD InStock
100496-418 100496-416
Earle’s balanced salt solutions (EBSS) Formula II
Buffers Cell Culture Buffers
All of our Earle’s Balanced Salt Solution contains: Potassium Chloride, Sodium Chloride, Sodium Phosphate Monobasic, and D-glucose. Some formulations may contain additional components. All of the solutions are prepared 1X strength.

Earle’s Balanced Salt Solutions Formula II contains:
KCl: 0.152 M
NaCl: 0.152 M
NaH₂PO₄·H₂O: 0.152 M
MgSO₄·7H₂O: 0.152 M
CaCl₂,·2H₂O: 0.152 M
NaHCO₃: 0.152 M
D-Glusose: 0.152 M
Phenol Red: 0.015 g/L

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty