Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
HANKS's Balanced Salts, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
11770-05 11768-10 11768-05 11769-10 11770-10 11769-05
100502-390EA 38.89 USD InStock
100502-390 100502-388 100496-426 100496-424 100502-386 100496-428
HANKS's Balanced Salts, Electron Microscopy Sciences
Buffers Cell Culture Buffers
All of our Hanks’ Balanced Salt Solutions contain: Potassium chloride, potassium phosphate monobasic, sodium chloride, sodium phosphate dibasic, and D-glucose. Some formulations may contain additional components. Use the table below as a quick reference for identifying the one that best suits your application.
All solutions are prepared in 1X strength.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty