Would you like to log in again?
Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
11772-10 11773-05 11773-10 11772-05 11774-10 11774-05
100503-018EA 61.77 USD InStock
100503-018 100496-432 100496-430 100496-434 100503-016 100503-014
Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences
Buffers Biological Buffers
All of our Dullbecco’s phosphate buffer saline solutions contain: potassium chloride, potassium phosphate monobasic, sodium chloride, sodium phosphate dibasic, and D-glucose.
  • Some formulations may contain additional components
  • Store at room temperature
Use the table below as a quick reference for identifying the one that best suits your application. All of the solutions are prepared 1X strength.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty