Would you like to log in again?
JB-4 Plus™ Embedding Kit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
JB-4™
14272-01 14272-00 14272-04 14272-06
100496-476EA 359.25 USD InStock
100496-476 100496-478 100491-338 100496-480
JB-4 Plus™ Embedding Kit, Electron Microscopy Sciences
Stains Microscopy and staining reagents
JB-4 Plus offers all of the same features as JB-4 except it produces less of an exothermic reaction than JB4 which is good for temperature sensitive tissues
JB-4 Plus Kit Consists of 500 mL Solution A , 15 mL Solution B , and 8 g Benzoyl Peroxide Catalyst.
It produces harder blocks which is ideal for dense samples, such as bone.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty