Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Formaldehyde/Zinc Fixative, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
15675-04 15675-01
100496-488EA 126.83 USD InStock
100496-488 100496-486
Formaldehyde/Zinc Fixative, Electron Microscopy Science
Stains Microscopy and staining reagents
Formaldehyde/zinc fixative is ready-to-use solution. Can be used with automated tissue processors.
  • Preserves fine nuclear details
  • Good for immunostaining
  • Contains no mercury
Works well with standard and special stains because zinc is a stain enhancing mordant.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty