Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formalin 10% (w/v), acetate buffered (neutral)

Supplier: Electron Microscopy Sciences
Warning
Synonyms: MethylaldehydeFormolParaformaldehyde aqueous solutionFormaldehyde

15738-01 15738-04
100496-498EA 51.73 USD InStock
100496-498 100496-500
Formalin 10% (w/v), acetate buffered (neutral)
Formalin
Buffered with sodium acetate in accordance with the A.F.I.P. manual of histology.

  • Filtered
  • Ready-to-use


Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: 100 °C (212 °F)
Density: 1 g/cm³ (8.345 lbs/gal.)
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 3334
ADR: 9,
Order Now
Specification Test Results
pH at 20°C 6.0 - 7.1

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty