Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formalin 10% (v/v), Phosphate buffered (Neutral)

Supplier: Electron Microscopy Sciences
Danger
Synonyms: MethylaldehydeFormolParaformaldehyde aqueous solutionFormaldehyde

15740 15740-04 15740-01
100503-922EA 41.89 USD InStock
100503-922 100496-502 100496-504
Formalin 10% (v/v), Phosphate buffered (Neutral)
Formalin
Buffered with phosphate in accordance with the A.f.I.P. manual of Histology.

  • Filtered
  • Ready-to-use


Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: 100 °C (212 °F)
Density: 0.98817 g/cm³ (8.246 lbs/gal.)
Flash Pt: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 1993
ADR: ,III
Order Now
Specification Test Results
pH at 25 °C 6.9 - 7.1

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty