Synonyms:
Methylaldehyde
, Formol
, Paraformaldehyde aqueous solution
, Formaldehyde
15741-04
15741-01
100500-246CS
442.31
USD
InStock
100500-246
100496-506
Formalin 10% in aqueous solution low odor, buffered
Formalin
10% formalin in Phosphate buffer according to the A.F.I.P. Manual of Histology.
This solution allows for long-term storage of tissue specimens.