Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Platinum, disc, drilled apertures 3 mm O.D., Thickness 0.12 mm

Supplier: Electron Microscopy Sciences
Synonyms: Platinium

43100-PT 43200-PT 43010-PT 43020-PT 43300-PT 43070-PT 43030-PT 43060-PT 43050-PT 43150-PT
100496-670EA 157.12 USD InStock
100496-670 100503-040 100496-662 100501-612 100496-654 100503-042 100496-666 100503-038 100496-658 100503-036
Platinum, disc, drilled apertures 3 mm O.D., Thickness 0.12 mm
Platinum
Platinum apertures are easily cleaned by either an aperture flamer or over a bunsen burner in the open air. We maintain a wide variety of Platinum Drilled Apertures in stock with different configurations suiting all of your needs.

Platinum Apertures size tolerances vary as follow:
• Nominal hole diameter: +/- 1 micron
• Disc diameter: +/- 10 microns
• Disc thickness: +/- 10 microns

Platinum Apertures size tolerances vary as follow:

Hole Sizes not listed are available upon special request.


Formula: Pt
MW: 195.08 g/mol
MDL Number: MFCD00011179
CAS Number: 7440-06-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty