Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Platinum, disc, drilled apertures 6.35 mm O.D., Thickness 0.06 mm

Supplier: Electron Microscopy Sciences
Synonyms: Platinium

47050-PT 47030-PT 47150-PT 47200-PT 47100-PT
100496-770EA 172.57 USD InStock
100496-770 100496-762 100496-760 100496-766 100496-768
Platinum, disc, drilled apertures 6.35 mm O.D., Thickness 0.06 mm
Platinum
Platinum apertures are easily cleaned by either an aperture flamer or over a bunsen burner in the open air. We maintain a wide variety of Platinum Drilled Apertures in stock with different configurations suiting all of your needs.

Platinum Apertures size tolerances vary as follow:
• Nominal hole diameter: +/- 1 micron
• Disc diameter: +/- 10 microns
• Disc thickness: +/- 10 microns

Platinum Apertures size tolerances vary as follow:

Hole Sizes not listed are available upon special request.


Formula: Pt
MW: 195.08 g/mol
MDL Number: MFCD00011179
CAS Number: 7440-06-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty