Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Micro-Mill® Grinder, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

50100-02 50100-01 50101-01
100496-818EA 4986.66 USD InStock
100496-818 100496-816 100496-814
Micro-Mill® Grinder, Electron Microscopy Sciences
Mills
Micro-Mill® Grinder can be used for fast, high-speed grinding of small samples from 20 to 50 mm (¾ to 2")

Micro-Mill® 1: With stainless steel blade, 0-5 minutes electrical timer with an ON position for continuous operation.
Micro-Mill® 2: With hard, stellate faced blade, with a momentary hold-down switch.

The unit comes with a stainless steel or hard-faced blade which rotates at speeds up to 12,000 rpm and will successfully grind bauxite, bone, ceramics, coal, Devorda's alloy, fertilizer, filter paper, fly ash, frozen tissue, glass, grain, kaolinites, leaves, limestone, muscle tissue, pharmaceuticals, pigment blends, plastics, rocks, roots, seeds, slags, soil, teeth, wood, etc. The grinding chamber measures diameter x depth: 54 x 25 mm (2 1/8 x 1"). Its overall dimensions are 14 x 18 x 28 cm (5 ½ x 7 x 11"). It comes with a 1.8 m (6 ft.)3-wire cord and grounded plug. Power: 115V, 60Hz or 220V, 60Hz. The 1/5 horsepower (150 watts) motor develops a torque of 30-ounce-inches (2.2 Kg-cm) at 12,000 rpm.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty