Would you like to log in again?
Tripod Polisher® TEM/SEM Sample Preparation, 590, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50116-50 50118-05 50116-60 50116-20 50116-65 50115-20 50116-31 50116-25 50116-70 50116-62 50116-40 50116-35 50116-36 50116-63 50116-15 50116-55 50116-30 50116-05 50116-75 50116-61 50116-64 50115-30 50115-10 50116-10
101411-584EA 2034.2 USD InStock
101411-584 100496-838 101411-588 101411-586 100502-430 100496-850 100496-840 100496-862 100491-340 100496-870 100502-428 100496-860 100496-854 100503-044 100496-844 100496-866 100496-842 100496-864 100496-858 100496-836 100496-848 100496-856 100496-846 100496-868
Tripod Polisher® TEM/SEM Sample Preparation, 590, Electron Microscopy Sciences
Polishers and Tumblers
A Tripod Polisher® was designed to prepare accurately micro sizes of TEM and SEM samples.
  • Limited ion milling
  • No strong chemicals are used to prepare the specimen
  • Reduce preferential thinning, radiation damage, and heating of the sample
Used to prepare both plan-view and cross-sections from a variety of sample materials, such as ceramics, composites, metals and, geological specimens. The same sample can be studied with both SEM and TEM and hence reduces sample preparation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty