Would you like to log in again?
Ion Mill Stage, Model 5901, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50118-01
100496-872EA 352.5 USD InStock
100496-872
Ion Mill Stage, Model 5901, Electron Microscopy Sciences
Specimen Mounting Supplies
The Ion Mill Stage is designed to accept the special SEM stub used with the Tripod Polisher®. The SEM stub is mounted to the Ion Mill Stage and the samples are briefly ion milled in order to remove fine scratches, polishing debris and to give the surface topography prior to SEM analysis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty