Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Chemical Polishing Fixture, 160, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50164-60
100497-034EA 1787.49 USD InStock
100497-034
Chemical Polishing Fixture, 160, Electron Microscopy Sciences
Polishers and Tumblers
The polishing of semiconductor materials often requires the use of harsh chemicals such as bromine or hydrofluoric acid solutions that would corrode the stainless steel from which most of the EMS fixtures are constructed
A special fixture for satisfying this requirement has been developed. The Model 160 is made of "PTFE" and is designed to hold crystal wafers samples up to 2 1/8" diameter while they are being polished with very corrosive chemicals. The sample is held in place by vacuum and the sample is polished by rubbing the exposed sample against a suitable polishing cloth soaked with the chemical solution. The Model 160 is ideal for damage free polishing of semiconductor materials with harsh chemicals and has been used extensively for polishing GaAs with bromine-methanol solutions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty