Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Diamond Lapping Film, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50354-20 50350-01 50351-10 50355-30 50351-30 50354-03 50354-35 50350-25 50355-01 50350-20 50355-25 50354-05 50351-20 50354-30 50351-25 50354-25 50351-05 50355-10 50354-10 50351-15 50355-35 50354-15 50351-03 50355-15 50350-03 50350-30 50355-05 50354-01 50351-35 50355-03 50350-10 50350-05 50355-20 50350-15 50351-01
100497-250EA 120.45 USD InStock
100497-250 100497-196 100497-230 100497-252 100497-198 100497-210 100497-254 100497-232 100497-234 100497-256 100497-236 100497-258 100497-214 100497-238 100497-216 100497-218 100497-260 100497-262 100497-240 100497-242 100497-220 100497-264 100497-244 100497-222 100497-266 100497-200 100497-224 100497-246 100497-202 100497-248 100497-226 100497-204 100497-228 100497-206 100497-208
Diamond Lapping Film, Electron Microscopy Sciences
Wipes Cleaning Kits and Pads
A selective diamond particle resin bonded to a uniform 3-mil polyester film backing. Excellent for polishing samples with differing hardness. Ideal for the preparation of cross sections for SEM/TEM analysis. Comes in 8" and 12" discs with plain back (P/B) or pressure sensitive adhesive back (PSA).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty