Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Glennel®, Diamond Suspension, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50372-11 50372-31 50372-10 50372-71 50372-40 50372-21 50372-60 50372-91 50372-30 50372-20 50372-81 50372-61 50372-80 50372-41 50372-90 50372-70
100497-360EA 134.53 USD InStock
100497-360 100497-350 100497-340 100497-362 100497-352 100497-342 100497-364 100497-354 100497-344 100497-366 100497-356 100497-368 100497-346 100497-358 100497-348 100497-338
Glennel®, Diamond Suspension, Electron Microscopy Sciences
Polishers and Tumblers
Glennel® Diamond Suspension assures a consistent, uniform viscosity and dispersion of diamond without any shaking or stirring.
  • A versatile, water based suspension designed for most production applications
  • Excellent lubrication
  • Uniform fast lapping action
  • Maximum cutting action
  • Compatible with all common extenders
The proprietary formulation guarantees that the particles will stay suspended indefinitely. It is ideal for automatic polishers and applications that require changing operators.
Packaging: All Diamond Suspensions are packed in 400 g bottles and have Medium Concentration prepared with Premium Polycrystalline Diamond in either Propylene Glycol (Oil based) or Water based formulas.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty