Would you like to log in again?
Glennel® Diamond Aerosol Spray, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50374-21 50374-16 50374-31 50374-06 50374-11 50374-10 50374-26
100497-392EA 132.21 USD InStock
100497-392 100497-372 100497-384 100497-374 100497-376 100497-388 100497-380
Glennel® Diamond Aerosol Spray, Electron Microscopy Sciences
Sprays
Glennel® Diamond Aerosol Spray provides unsurpassed cutting action and finish.
The inverted nozzle provides safe and easy access for difficult to reach areas while ensuring that every diamond particle is used. Special formulations allow the carrier to evaporate quickly, leaving a very uniform dispersion of diamond. This is especially useful when an operation requires a dry polishing cloth. Our Diamond Spray is available in 5 oz. cans.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty