Would you like to log in again?
Non-Marking Heel Grounder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60619-20 60619-10 60619-30
100497-450EA 17.03 USD InStock
100497-450 100497-452 100497-448
Non-Marking Heel Grounder, Electron Microscopy Sciences
Grounders Heel and Toe Grounders
E.Z. Close is your low cost logical choice heel grounder.
  • The Velcro® fastening system is adjust-able and comfortable
  • Charge decay: less than 0.01 sec.; 2.4×108 Ω
They are made from premium grade non-marking conductive rubber that is highly abrasion and wear resistant.
Certifications: Meets DOD-263A standards (USA).
Delivery information: Includes: 18" conductive tab and 1¹/₂", two-layer, rubber cup. Built in resistor.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty