Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Wheaton M-T Vial File® Storage Box for Vials, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
WHEATON®
60990-40 60990-46 60990-60 60990-66
100497-492EA 48.15 USD InStock
100497-492 100500-258 100500-260 100497-490
Wheaton M-T Vial File® Storage Box for Vials, Electron Microscopy Sciences
Boxes Boxes for Vials
This filing system offers a clean, compact, and convenient alternative for the storage of biological samples.
  • Clear polystyrene case, with hinged lid
  • Alpha-numerical index with individual partitions
Foam inserts are provided as well and they offer extra protection for the vials. Dimensions: 63/4L x 33/4W x 21/2H " (190 x 95 x 62 mm). M-T/40: stores up to 40 vials of 16 mm diameter x 50 mm height. M-T/60: stores up to 60 vials of 13 mm diameter x 50 mm height.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty