Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rotamix™ Rotators, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

61051-10 61050-10
100497-494EA 1154.93 USD InStock
100497-494 100497-496
Rotamix™ Rotators, Electron Microscopy Sciences
Shakers and Mixers Rotating/Rocking Shakers
The Rotamix units are designed with interchangeable 12" long rotary rods accommodating tubes ranging from 1.5 to 50 mL, which make the rotary universal.

  • Rotamix is made from steel, coated with a chemically resistant paint
  • Extremely quiet
  • Speed is variable from 0 to 80 rpm
  • All units are analog controlled
  • Suitable for use both in the cold room and the incubator

On the RKVSD, rotation speed is displayed on a large LCD.

Suitable applications:
- Immunoprecipitation
- Extractions and phase separations
- Bead mixing and extracting
- Blood mixing
- Bacterial cultures
- Embedding infiltration

Ordering information: Units come without racks. Racks are sold separately.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty