Would you like to log in again?
Thermo-Flask Stainless Steel, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61891-04 61891-02 61891-01
100497-510EA 1047.29 USD InStock
100497-510 100497-512 100497-514
Thermo-Flask Stainless Steel, Electron Microscopy Sciences
Dewars and Cryogenic Vessels
Thermo-Flask provides safe extended temperature retention for specimens stored in liquid nitrogen or solid CO2. The flasks features borosilicate glass inner liners which have been evacuate to 1 x 10 – 5 mm of Hg to provide maximum coolant retention.
  • Durable, corrosion-resistant stainless steel exterior construction
  • Carrying handle and vented lid secured by toggle clamps
  • Recommended for the safe transport of liquid nitrogen
  • Broad range of applications in industrial, educational and clinical fields
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty