Would you like to log in again?
Nalgene® Polyethylene Dewar Flask, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
62038-01 62038-02 62038-04 62038-10
100497-540EA 567.62 USD InStock
100497-540 100497-542 100503-050 100497-544
Nalgene® Polyethylene Dewar Flask, Electron Microscopy Sciences
Dewars and Cryogenic Vessels
High density polyethylene dewar flask with vented and insulated HDPE covers. Double walls are filled with foamed urethane to minimize heat exchange. May be used for short term storage of dry ice-solvent, liquid nitrogen, or for use as a hot bath. They are ideal for storing frozen biological specimens.
  • Moulded-in-ribs for stability
  • Carrying handle on the 1, 2, and 4 litre sizes
  • Unbreakable and chemically resistant
  • Withstand temps from –196 to +100 °C
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty