Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Digimatic Calipers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62062-20 62062-10
100497-546EA 355.25 USD InStock
100497-546 100497-548
Digimatic Calipers, Electron Microscopy Sciences
Calipers
All metal head with a plastic housing.
Provides a digital readout to 0.0005" / 0.01 mm for error-free reading and can measure steps. Function: zero setting, data hold, low battery alarm and data output. Provided with a data saving output port for connection with the SPC system. Powered by a single SR-44 battery. Battery life: approximately two years under normal use.
Specifications: Range: 0 – 6" / 0 - 150 mm; LCD resolution: 0.0005" / 0.0; Accuracy: ±0.001
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty