Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Micro-Tool Individual Micro Tools, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62091-15-25 62091-05-50 62091-02-25 62091-14 62091-06 62091-03-50 62091-15-50 62091-08-25 62091-25-25 62091-27-25 62091-26-12 62091-09-50 62091-28-50 62091-11-50 62091-23 62091-09-12 62091-02-50 62090-00 62091-12-50 62091-25-50 62091-16-0.50 62091-01-25 62091-02-0.12 62091-29-25 62091-24-025 62091-08-50 62091-18-50 62091-04-025 62091-28-25 62091-13-0.25 62091-01-50 62091-04-12 62091-27-50 62091-11-25 62091-03-12 62091-20-25 62091-26-50 62091-22 62091-10 62091-31 62091-24-12 62091-04-25 62091-29-50 62091-19 62091-25-12 62091-30 62091-32 62091-03-0.25 62091-07-0.50  62091-13-50 62091-27-12 62091-20-50 62091-04-50 62091-09-25 62091-26-25 62091-01-12 62091-12-25 62091-17 62091-24-25 62091-24-50 62091-18-25 62091-13-0.12 62091-05-25
100497-650EA 81.29 USD InStock
100497-650 100497-696 100501-658 100497-652 100497-730 100497-654 100497-698 100497-732 100502-468 100497-658 100501-656 100501-654 100502-470 100497-660 100501-648 100501-646 100497-664 100497-666 100497-700 100502-472 100497-668 100497-626 100497-704 100497-628 100497-706 102093-932 100497-708 102093-934 100501-644 102093-938 100501-642 100497-670 100501-638 100497-672 100501-636 100497-630 100497-676 100497-632 100497-710 100497-678 100497-712 100497-636 100497-714 100497-638 100497-716 100497-718 102093-924 100501-632 100501-628 100497-680 100497-682 100497-640 100497-684 100497-642 100497-686 100497-644 100497-646 100497-724 100497-648 100497-726 100497-728 100501-662 100501-660
Micro-Tool Individual Micro Tools, Electron Microscopy Sciences
Tool Sets
Micro-Tools are the smallest known precision tools available for the laboratory and micro electronics industry. These tools are designed for anyone who is engaged in microscope work, miniature applications as well as specimen manipulation.
  • Consists of a threaded handle (MT1), 4.75" long and an interchangeable threaded base with tool tip
  • Available in 32 different tip configurations with interchangeable handles
  • Tip material is hardened tool steel unless noted
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty