Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Standard Micro-Tool Sets, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62093-02 62093-04 62093-01 62093-03
100497-738EA 536.67 USD InStock
100497-738 100497-740 100497-742 100497-744
Standard Micro-Tool Sets, Electron Microscopy Sciences
Tool Sets
Micro-Needle Probe Set includes long Micro-needle, 90 degree Micro-needle, 45 degree Micro-needle, 90 degree Micro-hook, 180 degree Micro-hook, short Micro-needle, straight Ultra micro-needle, Micro-diamond-scriber, Micro-tool handle, and Micro-tool storage case.
Micro-Cutting Tool Set includes 3 sided Micro-graver, Micro-spade, oval Micro-graver, carbide Micro-scriber, Micro-tool storage case, Micro-chisel, Micro-knife 20 degree, Micro-scraper triangular, Micro-tool Handle.
Microscopist’s Set includes long Micro-needle, 20 degree Micro-knife, flexible Micro-spatula, 2-sided (metric & inch) Micro-scale, Micro-tool handle, Micro-chisel, Micro-diamond-scriber, 90 degree Micro-hook, straight Ultra-micro-needle, Micro-tool storage case.
Placement and Manipulating Set includes Micro-fork, 45 degree Micro-prober, Micro-spatula, 90 degree Micro-hook, Micro-tool handle, 10 mm Micro-scale, Micro-brush, Micro-manipulator, 30 degree bent Ultra-micro-needle, Micro-tool storage case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty